missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Rat, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP136PMI
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP136PMI 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP136PMI Supplier MilliporeSigma Supplier No. AP136P

Goat Polyclonal Antibody

Detect Rat IgG using this Goat anti-Rat IgG Antibody, HRP conjugate validated for use in ELISA, immunohistochemistry and western blotting.

Specifications

Antigen Goat anti-Rat IgG
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA. Reconstitute with 2mLof sterile distilled water.
Host Species Goat
Purification Method Affinity Purified
Quantity 2 mL
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C for up to 12 months.
Isotype IgG
Show More Show Less

For Research Use Only