missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rat IgG (H+L) Secondary Antibody, HRP, eBioscience™

Catalog No. 501129571
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501129571 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501129571 Supplier Invitrogen™ Supplier No. 18481882
Only null left

Goat Polyclonal Secondary Antibody

Description: This is affinity purified HRP-conjugated Goat Anti-Rat IgG (H+L). This antibody has minimal cross-reactivity to Mouse, Human, Bovine, Horse, and Rabbit Serum Proteins. Based on immunoelectrophoresis, the antibody reacts with the heavy chains on rat IgG and with thelight chains common to most rat immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. Applications Reported: HRP anti-rat IgG has been reported for use in immunoblotting (WB), immunohistochemical staining, and ELISA. Applications Tested: This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with mouse, human,bovine, horse, and rabbit serum proteins, but the antibody may cross-react with immunoglobμLins from other species.Suggested dilution ranges are:1:500 - 1:5,000 for immunohistochemistry on tissue sections 1:5,000 - 1:100,000 for ELISA and Western blotting with chromogenic substrates 1:10,000 - 1:200,000 for Western blotting with ECL substrates.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rat IgG (H+L)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate HRP
Formulation PBS with 50% glycerol and no preservative; pH 7.2
Host Species Goat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype Ig
Show More Show Less