missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rat IgG (H+L) Secondary Antibody, HRP

Catalog No. PI31470
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31470 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31470 Supplier Invitrogen™ Supplier No. 31470

Goat Polyclonal Secondary Antibody

This antibody has been successfully used in Western blot, IHC, ELISA and IP applications. Antibody Specificity: This antibody reacts with the heavy chains of rat IgG and with the light chains common to most rat immunoglobulins, based on immunoelectrophoresis. No antibody was detected against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 4°C until opened. Restore with 2.0 mL distilled water (0.8 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rat IgG (H+L)
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate HRP
Formulation PBS with 15mg/mL BSA and no preservative; pH 7.6
Host Species Goat
Purification Method Antigen affinity chromatography
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage 4°C
Product Type Secondary Antibody
Form Lyophilized
Isotype IgG
Show More Show Less