missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Rat, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
This Goat anti-Rat IgG Antibody, FITC conjugate, Species Adsorbed is validated for use in IF for the detection of Rat IgG.
Specifications
Specifications
| Antigen | Goat anti-Rat IgG Species adsorbed |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | FITC |
| Host Species | Goat |
| Quantity | 500 μg |
| Primary or Secondary | Secondary |
| Target Species | Rat |
For Research Use Only