missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Rat, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP136FMI
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP136FMI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP136FMI Supplier MilliporeSigma Supplier No. AP136F
Only null left

Goat Polyclonal Antibody

Goat anti-Rat IgG Antibody, FITC conjugate is an antibody against Rat IgG for use in immunofluoresence.

Specifications

Antigen Goat anti-Rat IgG
Applications Immunofluorescence
Classification Polyclonal
Concentration 26mg/mL
Conjugate FITC
Host Species Goat
Immunogen Highly purified Rat IgG isolated from pooled normal rat serum.
Quantity 2 mg
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C protected from light.

DO NOT FREEZE.
Show More Show Less

For Research Use Only