missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Rat, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Goat anti-Rat IgG Antibody, Cy3 conjugate detects level of Rat IgG and has been published and validated for use in immunofluoresence.
Specifications
Specifications
| Antigen | Goat anti-Rat IgG |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Cyanine3 |
| Host Species | Goat |
| Quantity | 2 mg |
| Primary or Secondary | Secondary |
| Target Species | Rat |
For Research Use Only