missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Rat, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP136AMI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP136AMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP136AMI Supplier MilliporeSigma Supplier No. AP136A
Only null left

Goat Polyclonal Antibody

Goat anti-Rat IgG Antibody, Alkaline Phosphatase conjugate is an antibody against Rat IgG for use in ELISA and western blotting.

Specifications

Antigen Goat anti-Rat IgG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.6mg/mL
Conjugate Alkaline Phosphatase (AP)
Formulation RECONSTITUTION:

Reconstitute with sterile distilled water to match the volume indicated on the vial label. Centrifuge or vortex this product if it is not completely clear after standing for 1-2 hours at room temperature.
Host Species Goat
Immunogen Highly purified Rat IgG isolated from pooled normal rat serum.
Quantity 1 mL
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C lyophilized until opened. DO NOT FREEZE.
Form Lyophilized
Show More Show Less

For Research Use Only