missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat Albumin Polyclonal Antibody, MP Biomedicals
Supplier: MP Biomedicals Inc 0855710
Description
Host Species: Goat
Species Reactivity: Rat
Specifications
| Albumin | |
| Unconjugated | |
| 2 mL | |
| Rat |
| Polyclonal | |
| Goat | |
| Primary | |
| Lyophilized |