missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Rabbit, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™
Goat Polyclonal Antibody
Supplier: MilliporeSigma AP187R
Description
Detect Rabbit IgG using this Goat anti-Rabbit IgG Antibody, Rhodamine conjugate, Species Adsorbed validated for use in immunofluoresence.Specifications
| Goat anti-Rabbit IgG Species adsorbed | |
| Polyclonal | |
| Rhodamine | |
| Goat | |
| Secondary | |
| 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. |
| Immunofluorescence | |
| 1.5mg/mL | |
| Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, 0.01% Thimerosal and 0.05 Sodium Azide. RECONSTITUTION: Reconstitute to 1mg/mL with sterile distilled water. | |
| 1.5 mg | |
| Rabbit | |
| Lyophilized |
For Research Use Only