missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Rabbit, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP132RMI
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP132RMI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP132RMI Supplier MilliporeSigma Supplier No. AP132R

Goat Polyclonal Antibody

Goat anti-Rabbit IgG Antibody, Rhodamine conjugate is an antibody against Rabbit IgG for use in immunofluoresence.

Specifications

Antigen Goat anti-Rabbit IgG
Applications Immunofluorescence
Classification Polyclonal
Conjugate Rhodamine
Formulation Lyophilized. Buffer = 0.02M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, and 0.1% sodium azide. RECONSTITUTION: Reconstitute with 2mLof sterile distilled water RHODAMINE/PROTEIN: Approximately 0.62 (A550/A280).
Host Species Goat
Quantity 2 mg
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only