missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Rabbit, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP132MI
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP132MI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP132MI Supplier MilliporeSigma Supplier No. AP132
Only null left

Goat Polyclonal Antibody

Goat anti-Rabbit IgG Antibody is an antibody against Rabbit IgG for use in ELISA, immunoprecipitation and western blotting.

Specifications

Antigen Goat anti-Rabbit IgG
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 2mg/mL
Conjugate Unconjugated
Host Species Goat
Immunogen Highly purified Rabbit IgG isolated from pooled normal Rabbit Serum.
Quantity 2 mg
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C
Form Purified
Show More Show Less

For Research Use Only