missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Rabbit, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP132PMI
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP132PMI 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP132PMI Supplier MilliporeSigma Supplier No. AP132P
Only null left

Goat Polyclonal Antibody

This Goat anti-Rabbit IgG Antibody, Peroxidase Conjugated is validated for use in ELISA, immunohistochemistry, western blotting for the detection of Rabbit IgG.

Specifications

Antigen Goat anti-Rabbit IgG
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate HRP
Formulation Purified goat IgG conjugated to horseradish peroxidase in buffer containing 0.01M Phosphate Buffered Saline, pH 7.1 with 15mg/mL BSA and 0.01% Thimerosal. Lyophilized. RECONSTITUTION: Reconstitute with sterile distilled water to match the volume indicated on the label.
Host Species Goat
Purification Method Affinity Purified
Quantity 2 mL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C for up to 12 months.
Isotype IgG
Show More Show Less

For Research Use Only