missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Rabbit, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Goat Anti-Rabbit IgG Antibody, HRP-conjugate detects level of Rabbit IgG and has been published and validated for use in western blot, immunohistochemistry, ELISA.
Specifications
Specifications
| Antigen | Goat anti-Rabbit IgG |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | HRP |
| Formulation | Goat polyclonal conjugated with horseradish peroxidase IgG in buffer containing 50% storage buffer 0.02M potassium phosphate, 0.15M NaCl, pH 7.2, 10mg/mL BSA, 0.01% (w/v) gentamicin sulfate and 50% glycerol. Rehydration: Add 500 μL sterile, distilled water containing 50% glycerol, to make a 1 mg/mL stock solution. |
| Host Species | Goat |
| Immunogen | Highly purified whole rabbit IgG. |
| Quantity | 500 μg |
| Primary or Secondary | Secondary |
| Show More |
For Research Use Only