Learn More
Goat anti-Rabbit IgG (Heavy chain), Superclonal™ Recombinant Secondary Antibody, HRP, Invitrogen™
Goat Recombinant Polyclonal Secondary Antibody
Supplier: Invitrogen LSA27036
Description
The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig and donkey IgG is observed. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
| Rabbit IgG (Heavy chain) | |
| Polyclonal | |
| HRP Stabilizer, PBS with proprietary preservative | |
| Recombinant full-length protein | |
| RUO | |
| Rabbit | |
| Liquid |
| ELISA, Immunoprecipitation, Peptide Array, Western Blot | |
| HRP | |
| Goat | |
| Gravity column chromatography | |
| Secondary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| IgG |