missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rabbit IgG (Heavy chain) Superclonal™ Secondary Antibody, Alexa Fluor™ Plus 405

Catalog No. A55052
Encompass
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55052 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A55052 Supplier Invitrogen™ Supplier No. A55052

Goat Recombinant Superclonal Secondary Antibody

Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity and superior lot-to-lot consistency. Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig and donkey IgG is observed. Obtain superior images with the new Invitrogen Alexa Fluor Plus secondary antibodies. Make your low abundant targets visible, minimize time spent optimizing, and make your precious samples count. Alexa Fluor Plus secondary antibodies provide brighter signal compared to leading Alexa Fluor secondary antibodies, providing you with better signal-to-noise for your critical experiments. Alexa Fluor Plus secondary antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor Plus secondary antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning images. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically.

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rabbit IgG (Heavy chain)
Applications Flow Cytometry, Immunocytochemistry
Classification Recombinant Superclonal
Concentration 2 mg/mL
Conjugate Alexa Fluor Plus 405
Formulation Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.5
Host Species Goat
Immunogen Purified Rabbit Immunoglobulin.
Purification Method Affinity chromatography
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less