missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rabbit IgG (Heavy chain), Superclonal™ Recombinant Secondary Antibody, Alexa Fluor™ 555, Invitrogen™

Catalog No. A27039
Encompass
Catalog No. A27039 Supplier Thermo Fisher Scientific Supplier No. LSA27039
Only null left

Goat Recombinant Polyclonal Secondary Antibody

The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig and donkey IgG is observed. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rabbit IgG (Heavy chain)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 555
Formulation PBS with 5mM sodium azide
Host Species Goat
Immunogen Recombinant full-length protein
Purification Method Gravity column chromatography
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 4° C, store in dark, DO NOT FREEZE!
Form Liquid
Isotype IgG
Show More Show Less