Learn More
Invitrogen™ Goat anti-Rabbit IgG (H+L) Secondary Antibody, AP
Description
This antibody has been successfully used in Western blot, IHC and IP applications. Antibody Specificity: Based on immunoelectrophoresis, this antibody reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 4°C until opened. Restore with 1.0 mL distilled water (0.6 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.
Specifications
Specifications
| Antigen | Rabbit IgG (H+L) |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Alkaline Phosphatase (AP) |
| Formulation | TBS with 15mg/mL BSA and no preservative; pH 8 |
| Host Species | Goat |
| Immunogen | Purified Rabbit IgG, whole molecule. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mL |
| Show More |