missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Rabbit, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP307RMI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP307RMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP307RMI Supplier MilliporeSigma Supplier No. AP307R

Goat Polyclonal Antibody

This Goat anti-Rabbit IgG Antibody, (H+L) Rhodamine conjugate is validated for use in IF for the detection of Rabbit IgG.

Specifications

Antigen Goat anti-Rabbit IgG
Applications Immunofluorescence
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Rhodamine
Formulation F/P MOLAR RATIO: 2-3
Host Species Goat
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C in undiluted format for 12 months.
Show More Show Less

For Research Use Only