missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG (H+L) Goat anti-Mouse, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™
Goat Polyclonal Antibody
Supplier: MilliporeSigma 12510
Description
Goat anti-Rabbit IgG (H+L) Antibody, Rhodamine conjugate detects level of Rabbit IgG (H+L) and has been published and validated for use in flow cytometry, immunofluoresence, immunocytochemistry, immunohistochemistry.Specifications
| Goat anti-Rabbit IgG (HL) | |
| Polyclonal | |
| 0.02M potassium phosphatase, pH 7.2, 150mM NaCl, 0.01% sodium azide and 10mg/mL BSA | |
| 500 μg | |
| Mouse | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| Rhodamine | |
| Goat | |
| Secondary | |
| Lyophilized: 2 years at 4°C; Rehydrated: −20°C |
For Research Use Only