missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rabbit IgG (H+L) ReadyProbes™ Secondary Antibody, Biotin

Catalog No. SA510419
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA510419 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA510419 Supplier Invitrogen™ Supplier No. SA510419

Goat Polyclonal Secondary Antibody

Recognizes both heavy and light chains (H+L). No pipetting or dilutions required. Just tip and drip enough reagent to adequately cover the tissue or cells (typically 2-3 drops).

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rabbit IgG (H+L)
Applications ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Not Determined
Conjugate Biotin
Formulation PBS with carrier protein and 0.08% sodium azide
Host Species Goat
Purification Method Affinity chromatography
Quantity 50 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less