missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Rabbit, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP307PMI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP307PMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP307PMI Supplier MilliporeSigma Supplier No. AP307P
Only null left

Goat Polyclonal Antibody

Detect Rabbit IgG using this Goat anti-Rabbit IgG Antibody, (H+L) HRP conjugate validated for use in ELISA, immunocytochemistry, immunohistochemistry and western blotting.

Specifications

Antigen Goat anti-Rabbit IgG
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate HRP
Formulation Purified Goat IgG in buffer containing liquid in PBS, pH 7.1 with 15mg/mL BSA and 0.01% thimerosal.
Host Species Goat
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C in undiluted aliquots for upto 12 months.
Isotype IgG
Show More Show Less

For Research Use Only