missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, HRP

Goat Polyclonal Secondary Antibody

$438.00

Specifications

Antigen Rabbit IgG (H+L) Highly Cross-Adsorbed
Concentration 1 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
A16110
Encompass
View Documents Promotion Details
Invitrogen™
A16110
1 mg
Each for $438.00
Only null left
 
Description

Description

Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. Store lyophilized material at 2-8 °C. For long term storage after reconstitution, dilute with 50% glycerol and store at -20 °C as a liquid. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin rabbit serum proteins, serum proteins from bovine, goat, human, mouse, or rat, IgG from bovine, goat, human, mouse or rat.

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Rabbit IgG (H+L) Highly Cross-Adsorbed
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Lyophilized
IgG
Rabbit
Gamma Immunoglobins Heavy and Light chains.
Secondary Antibody
1 mg/mL
ELISA, Immunohistochemistry, Western Blot
HRP
Goat
RUO
PBS with 1% BSA and 0.1% ProClin 150; pH 7.2
Secondary
Affinity chromatography
Videos
SDS
Documents

Documents