missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG (H+L) Goat anti-Rabbit, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Detect Rabbit IgG (H+L) using this Goat anti-Rabbit IgG (H+L) Antibody, FITC conjugate validated for use in flow cytometry, immunofluoresence, immunocytochemistry, immunohistochemistry.
Specifications
Specifications
| Antigen | Goat anti-Rabbit IgG (HL) |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | Lyophilized powder |
| Host Species | Goat |
| Quantity | 500 μg |
| Primary or Secondary | Secondary |
| Target Species | Rabbit |
| Content And Storage | 4°C for up to 12 months. |
| Show More |
For Research Use Only