Learn More
Invitrogen™ Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP
Goat Polyclonal Secondary Antibody
$435.00
Specifications
| Antigen | Rabbit IgG (H+L) Cross-Adsorbed |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
Description
Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. Store lyophilized material at 2-8 °C. For long term storage after reconstitution, dilute with 50% glycerol and store at -20 °C as a liquid. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin rabbit serum proteins, serum proteins from bovine, human, or mouse , IgG from human or mouse. Based on IEP, this antibody reacts with: heavy (gamma) chains on rabbit IgG; light chains on all rabbit immunoglobulins. Affinity purified antibody is > 95% based on SDS-PAGE.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
| Rabbit IgG (H+L) Cross-Adsorbed | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Lyophilized | |
| IgG | |
| Rabbit | |
| Purified Rabbit IgG, whole molecule. | |
| Secondary Antibody |
| 1 mg/mL | |
| ELISA, Immunohistochemistry, Western Blot | |
| HRP | |
| Goat | |
| RUO | |
| PBS with 1% BSA and 0.1% ProClin 150; pH 7.2 | |
| Secondary | |
| Affinity chromatography |