missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Rabbit IgG (H+L) Cross-Adsorbed ReadyProbes™ Secondary Antibody, Alexa Fluor™ 594
Goat Polyclonal Secondary Antibody
Supplier: Invitrogen™ R37117
Description
No pipetting or dilutions required. Just tip and drip two drops per mL buffer to stain your cells.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Chemical Identifiers
| Rabbit IgG (H+L) Cross-Adsorbed | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Alexa Fluor 594 | |
| Goat | |
| RUO | |
| PBS with 0.1% BSA and <0.1% sodium azide | |
| Secondary | |
| Affinity chromatography |
| 4°C, store in dark | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Rabbit | |
| Gamma Immunoglobin. | |
| Secondary Antibody |
Specifications
| Rabbit IgG (H+L) Cross-Adsorbed | |
| Polyclonal | |
| PBS with 0.1% BSA and <0.1% sodium azide | |
| Gamma Immunoglobin. | |
| 2 x 2.5 mL | |
| Secondary | |
| 4°C, store in dark | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Alexa Fluor 594 | |
| Goat | |
| Affinity chromatography | |
| RUO | |
| Rabbit | |
| Secondary Antibody | |
| IgG |