missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Rabbit, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP156PMI
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP156PMI 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP156PMI Supplier MilliporeSigma Supplier No. AP156P
Only null left

Goat Polyclonal Antibody

This Goat anti-Rabbit IgG Antibody, flow cytometry, HRP conjugate is validated for use in ELISA, western blotting for the detection of Rabbit IgG.

Specifications

Antigen Goat anti-Rabbit IgG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.8mg/mL
Conjugate HRP
Formulation Liquid. Buffer =0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, and mixed with 15mg/mL BSA as stabilizer containing no preservatives. RECONSTITUTION: Reconstitute in 2mL deionized water.
Host Species Goat
Quantity 2 mL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Liquid
Show More Show Less

For Research Use Only