missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Rabbit, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Detect Rabbit IgG using this Goat anti-Rabbit IgG Antibody, F(ab′)2, biotin-SP conjugate validated for use in ELISA and western blotting.
Specifications
Specifications
| Antigen | Goat anti-Rabbit IgG |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Host Species | Goat |
| Quantity | 500 μL |
| Primary or Secondary | Secondary |
| Target Species | Rabbit |
For Research Use Only