missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Rabbit, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
This Goat anti-Rabbit IgG Antibody, Cy3 conjugate is validated for use in IF for the detection of Rabbit IgG.
Specifications
Specifications
| Antigen | Goat anti-Rabbit IgG |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Cyanine3 |
| Formulation | Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, and 0.1% sodium azide. RECONSTITUTION: Reconstitute with 2mLof sterile distilled water |
| Host Species | Goat |
| Quantity | 2 mg |
| Primary or Secondary | Secondary |
| Target Species | Rabbit |
| Content And Storage | 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. |
| Show More |
For Research Use Only