missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Rabbit, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
This Goat anti-Rabbit IgG Antibody, biotin-SP conjugate, Species Adsorbed is validated for use in ELISA, western blotting for the detection of Rabbit IgG.
Specifications
Specifications
| Antigen | Goat anti-Rabbit IgG Species adsorbed |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.1mg/mL |
| Conjugate | Biotin |
| Formulation | Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, 0.01% Thimerosal and 0.05% Sodium Azide RECONSTITUTION: Reconstitute with 1.5mLsterile distilled water. |
| Host Species | Goat |
| Quantity | 1.5 mL |
| Primary or Secondary | Secondary |
| Target Species | Rabbit |
| Show More |
For Research Use Only