missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit Complement C3 Polyclonal Anitbody, FITC, MP Biomedicals
Description
Host Species: Goat
Species Reactivity: Rabbit
Specifications
Specifications
| Antigen | Complement C3 |
| Classification | Polyclonal |
| Conjugate | FITC |
| Host Species | Goat |
| Purification Method | Affinity Purified |
| Quantity | 2 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Rabbit |
| Product Type | Antibody |
| Show More |