missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Light chain Goat anti-Mouse, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
This Goat anti-Mouse light chain Antibody, Cy3 conjugate is validated for use in ELISA, western blotting for the detection of Mouse light chain.
Specifications
Specifications
| Antigen | Goat anti-Mouse light chain |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Cyanine3 |
| Formulation | Lyophilized powder from a solution with 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 0.05% Sodium Azide. 15mg/mL BSA as stabilizer. Purified by immunoaffinity chromatography. |
| Host Species | Goat |
| Immunogen | purified mouse IgG light chain. |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Primary or Secondary | Secondary |
| Show More |
For Research Use Only