missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Light chain Goat anti-Mouse, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Goat anti-Mouse light chain Antibody, Alkaline Phosphatase conjugate detects level of Mouse light chain and has been published and validated for use in ELISA and western blotting.
Specifications
Specifications
| Antigen | Goat anti-Mouse light chain |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alkaline Phosphatase (AP) |
| Formulation | Lyophilized from a solution in 0.01M Tris-HCl, 0.25M NaCl, pH 8.0, 0.05% Sodium Azide. 15mg/mL BSA as stabilizer.Purified by immunoaffinity chromatography. |
| Host Species | Goat |
| Immunogen | purified mouse IgG light chain. |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Primary or Secondary | Secondary |
| Show More |
For Research Use Only