missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM+IgG+IgA (H+L) Goat anti-Mouse, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Detect Mouse IgM+IgG+IgA (H+L) using this Goat anti-Mouse IgM+IgG+IgA (H+L) Antibody, Species Adsorbed validated for use in ELISA.
Specifications
Specifications
| Antigen | Goat anti-Mouse IgM IgG IgA (HL) Species adsorbed |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified by affinity chromatography on pooled mouse IgM+IgG+IgA covalently linked to agarose. Liquid in 2.0mLof 100mM borate buffered saline, pH 8.2. |
| Host Species | Goat |
| Immunogen | pooled antisera from goats hyperimmunized with mouse IgM, IgG and IgA paraproteins. |
| Purification Method | Affinity Purified |
| Quantity | 2 mg |
| Primary or Secondary | Secondary |
| Show More |
For Research Use Only