missing translation for 'onlineSavingsMsg'
Learn More

IgM+IgG+IgA Goat anti-Mouse, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ502BMI
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ502BMI 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ502BMI Supplier MilliporeSigma Supplier No. AQ502B
Only null left

Goat Polyclonal Antibody

Goat anti-Mouse IgM+IgG+IgA Antibody, F(ab′)2, biotin conjugate, Species Adsorbed detects level of Mouse IgM+IgG+IgA and has been published and validated for use in ELISA and immunofluoresence.

Specifications

Antigen Goat anti-Mouse IgM IgG IgA F(ab')2 Species adsorbed
Applications ELISA, Immunofluorescence
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Purified by gel filtration chromatography of pepsin digested antibody. Liquid in 1.0mLPBS/NaN3.
Host Species Goat
Immunogen pepsin digest of mouse Ig (H, L).
Purification Method Gel filtration chromatography
Quantity 0.5 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C under sterile conditions for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only