missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 568
Description
Product will be shipped at Room Temperature.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Mouse IgM (Heavy chain) Cross-Adsorbed |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Alexa Fluor 568 |
| Formulation | PBS with 5mM sodium azide; pH 7.5 |
| Host Species | Goat |
| Immunogen | Mouse Mu immunoglobulin. |
| Purification Method | Purified |
| Quantity | 500 μg |
| Show More |
Frequently Asked Questions (FAQs)
I have a goat anti-mouse Alexa Fluor anti-IgM secondary antibody. Will it label my IgG primary antibody as well?
Not likely. With any secondary antibody, you need to consider what other isotypes and species they are cross-adsorbed against. If it is cross-adsorbed against other species or specific chains, cross-reactivity should be minimal or none. Our goat anti-IgM secondary antibodies have been cross-adsorbed against IgG and therefore should exhibit minimal binding to IgG.