missing translation for 'onlineSavingsMsg'
Learn More

IgM μ chain Goat anti-Mouse, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP500MI
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP500MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP500MI Supplier MilliporeSigma Supplier No. AP500

Goat Polyclonal Antibody

This Goat anti-Mouse IgM μ chain Antibody, Species Adsorbed is validated for use in ELISA for the detection of Mouse IgM μ chain.

Specifications

Antigen Goat anti-Mouse IgM μ chain Species adsorbed
Applications ELISA
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified by affinity chromatography on mouse IgM covalently linked to agarose. Liquid in 1.0mLof 100mM borate buffered saline, pH 8.2.
Host Species Goat
Immunogen purified mouse IgM μ chain. Pooled antisera from goats hyperimmunized with mouse IgM paraproteins.
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C under sterile conditions for up to 12 months
Show More Show Less

For Research Use Only