missing translation for 'onlineSavingsMsg'
Learn More

IgM Goat anti-Mouse, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP128RMI
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP128RMI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP128RMI Supplier MilliporeSigma Supplier No. AP128R
Only null left

Goat Polyclonal Antibody

Goat anti-Mouse IgM Antibody, μ chain, Rhodamine conjugate is an antibody against Mouse IgM for use in immunofluoresence.

Specifications

Antigen Goat anti-Mouse IgM
Applications Immunofluorescence
Classification Polyclonal
Conjugate Rhodamine
Host Species Goat
Quantity 2 mg
Primary or Secondary Secondary
Target Species Mouse

For Research Use Only