missing translation for 'onlineSavingsMsg'
Learn More

IgM μ chain Goat anti-Mouse, Cy5, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP500SMI
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP500SMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP500SMI Supplier MilliporeSigma Supplier No. AP500S

Goat Polyclonal Antibody

Goat anti-Mouse IgM μ chain Antibody, Cy5 conjugate, Species Adsorbed detects level of Mouse IgM μ chain and has been published and validated for use in ELISA and immunofluoresence.

Specifications

Antigen Goat anti-Mouse IgM μ chain Species adsorbed
Applications ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1mg/mL
Conjugate Cyanine5
Formulation Purified by affinity chromatography on mouse IgM covalently linked to agarose. Liquid in 1.0mLPBS/NaN3.
Host Species Goat
Immunogen mouse IgM μ chain. Pooled antisera from goats hyperimmunized with mouse IgM paraproteins.
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C under sterile conditions for up to 12 months. Avoid repeated freeze/thaw cycles. DO NOT FREEZE.
Show More Show Less

For Research Use Only