missing translation for 'onlineSavingsMsg'
Learn More

IgM μ chain Goat anti-Mouse, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP500AMI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP500AMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP500AMI Supplier MilliporeSigma Supplier No. AP500A

Goat Polyclonal Antibody

Goat anti-Mouse IgM μ chain Antibody, Alkaline Phosphatase conjugate, Species Adsorbed detects level of Mouse IgM μ chain and has been published and validated for use in ELISA.

Specifications

Antigen Goat anti-Mouse IgM μ chain Species adsorbed
Applications ELISA
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation Purified by affinity chromatography on mouse IgM covalently linked to agarose. Liquid in 50mM Tris/1mM MgCl2/50% Glycerol, pH 8.0, containing 0.1% NaN3 as preservative.
Host Species Goat
Immunogen purified mouse IgM μ chain. Pooled antisera from goats hyperimmunized with mouse IgM paraproteins.
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C under sterile conditions for up to 12 months −20°C in undiluted aliquots for long term storage. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only