missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Mouse IgG2b Secondary Antibody, Alexa Fluor™ 633
Goat Polyclonal Secondary Antibody
Supplier: Invitrogen™ A21146
Description
Fluorescence of this long-wavelength Alexa Fluor dye is not visible by looking through a conventional fluorescence microscope. Product will be shipped at Room Temperature.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Chemical Identifiers
| Mouse IgG2b | |
| 4°C, store in dark | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Mouse IgG2b. | |
| Secondary Antibody |
| 2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Alexa Fluor 633 | |
| Goat | |
| RUO | |
| PBS with 5mM sodium azide; pH 7.5 | |
| Secondary | |
| Purified |
Specifications
| Mouse IgG2b | |
| Polyclonal | |
| Alexa Fluor 633 | |
| Goat | |
| Purified | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 5mM sodium azide; pH 7.5 | |
| Mouse IgG2b. | |
| 500 μg | |
| Secondary | |
| 4°C, store in dark | |
| Liquid |