missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Mouse IgG2a Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 594

Catalog No. a21135
Encompass
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21135 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21135 Supplier Invitrogen™ Supplier No. A21135

Goat Polyclonal Secondary Antibody

Product will be shipped at Room Temperature.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Mouse IgG2a Cross-Adsorbed
Applications Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Alexa Fluor 594
Formulation PBS with 5mM sodium azide; pH 7.5
Host Species Goat
Immunogen Mouse IgG2a.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C, store in dark
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Do you offer an alternative for the discontinued 3-Germ Layer ICC Kit (Cat. No. A25538)?

We do not have a direct alternative for the discontinued 3-Germ Layer ICC Kit (Cat. No. A25538). However, we do have alternative primary and secondary antibodies, as well as reagents, that can be used for trilineage differentiation. Please note that we have not internally validated the use of all these reagents together.

We recommend the following primary antibodies:

Alpha-Smooth Muscle Actin Monoclonal Antibody (1A4 (asm-1)) (Cat. No. MA5-11547)

alpha-Fetoprotein Monoclonal Antibody (AFP3) (Cat. No. 14-6583-80)

beta-3 Tubulin Monoclonal Antibody (2G10) (Cat. No. MA1-118)

We recommend the following secondary antibodies:

Goat anti-Mouse IgG2a Cross-Adsorbed, Alexa Fluor 555 (Cat. No. A21137)

Goat anti-Mouse IgG2a Cross-Adsorbed, Alexa Fluor 594 (Cat. No. A21135)

https://www.thermofisher.com/antibody/product/Goat-anti-Mouse-IgG1-Cross-Adsorbed-Secondary-Antibody-Polyclonal/A-21121

We recommend using the following reagents:

https://www.thermofisher.com/order/catalog/product/88-8824-00

NucBlue Fixed Cell ReadyProbes Reagent (DAPI) (Cat. No. R37606)

Blocker BSA (Cat. No. 37520)

DPBS (10X), no calcium, no magnesium (Cat. No. 14200075)

I have two primary antibodies from the same host species that I wish to use to label the same cells. How do I choose secondary antibodies?

If the two primary antibodies are from different isotypes (such as mouse IgG1 and IgG2a), there are isotype-specific secondary antibodies to choose from. The other option is to make direct conjugates of the primary antibodies through the use of antibody label kits, reactive dyes, or custom conjugations.