missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG, Whole Molecule Polyclonal Secondary Antibody, MP Biomedicals
Supplier: MP Biomedicals Inc 0855455
Description
Host Species: Goat
Isotype: IgG
Species Reactivity: Mouse
Specifications
| IgG, Whole Molecule | |
| Total Protein: 24.6 mg/mL; Antibody Protein: 10 mg/mL | |
| Goat | |
| Secondary | |
| Lyophilized |
| Polyclonal | |
| Unconjugated | |
| 2 mL | |
| Mouse | |
| IgG |