missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Mouse, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP181RMI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP181RMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP181RMI Supplier MilliporeSigma Supplier No. AP181R

Goat Polyclonal Antibody

Goat anti-Mouse IgG Antibody, Rhodamine conjugate, Species Adsorbed detects level of Mouse IgG and has been published and validated for use in immunofluoresence.

Specifications

Antigen Goat anti-Mouse IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Concentration 1.4mg/mL
Conjugate Rhodamine
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6. RECONSTITUTION: Reconstitute to 1mg/mL with sterile distilled water.
Host Species Goat
Quantity 500 μg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only