missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Mouse Lambda Light Chain Secondary Antibody, AP

Catalog No. PIPA186022
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA186022 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA186022 Supplier Invitrogen™ Supplier No. PA186022

Goat Polyclonal Secondary Antibody

Goat anti-Mouse Lambda Light Chain Secondary Antibody for ELISA

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Mouse Lambda Light Chain
Applications ELISA
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation TBS with 50% glycerol, 1mM MgCl2 and <0.1% sodium azide
Host Species Goat
Immunogen Murine paraproteins with lambda 1 and lambda 2 light chains.
Purification Method Affinity chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less