missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Mouse, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. 12349MI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12349MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12349MI Supplier MilliporeSigma Supplier No. 12349
Only null left

Goat Polyclonal Antibody

Goat Anti-Mouse IgG Antibody, HRP conjugate is an antibody against Mouse IgG for use in ELISA, immunohistochemistry and western blotting.

Specifications

Antigen Goat anti-Mouse IgG
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation Purified goat polyclonal IgG in buffer containing conjugated to horseradish peroxidase lyophilized from 0.02M Potassium Phosphate, 0.15M NaCl, pH 7.2, 10mg/mL BSA, and 0.01% gentamicin sulfate. Rehydration: Add 500 μL sterile, distilled water containing 50% glycerol, to make a 1 mg/mL stock solution
Rehydration: Add 500μL sterile, distilled water conta
Host Species Goat
Immunogen Highly purified whole mouse IgG.
Quantity 500 μg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C for up to 12 months
Form Purified
Show More Show Less

For Research Use Only