Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody
Goat Recombinant Superclonal Secondary Antibody
Supplier: Invitrogen™ A28174
Description
The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with rabbit, rat, human, bovine, guinea pig and donkey IgG is observed. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Chemical Identifiers
| Mouse IgG (H+L) | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Recombinant Superclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Recombinant full-length protein. | |
| Secondary Antibody |
| 1 mg/mL | |
| ELISA, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Goat | |
| RUO | |
| PBS with 0.09% sodium azide | |
| Secondary | |
| Protein G |
Specifications
| Mouse IgG (H+L) | |
| Recombinant Superclonal | |
| Unconjugated | |
| Goat | |
| Protein G | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Recombinant full-length protein. | |
| 1 mg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |