missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Secondary Antibody, AP
Goat Polyclonal Secondary Antibody
Supplier: Invitrogen™ PA128755
Description
PA1-28755 detects Mouse IgG from mouse samples.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Chemical Identifiers
| Mouse IgG (H+L) | |
| 4°C, do not freeze | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Mouse IgG whole molecule. | |
| Secondary Antibody |
| 1 mg/mL | |
| Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
| Alkaline Phosphatase (AP) | |
| Goat | |
| RUO | |
| TBS with 0.001M MgCl2, 0.0001MM ZnCl2, 50% glycerol, 1% BSA and 0.09% sodium azide; pH 8 | |
| Secondary | |
| Antigen affinity chromatography |
Specifications
| Mouse IgG (H+L) | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
| Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| TBS with 0.001M MgCl2, 0.0001MM ZnCl2, 50% glycerol, 1% BSA and 0.09% sodium azide; pH 8 | |
| Mouse IgG whole molecule. | |
| 100 μg | |
| Secondary | |
| 4°C, do not freeze | |
| Liquid |