missing translation for 'onlineSavingsMsg'
Learn More

IgG (H+L) Goat anti-Mouse, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. 12509MI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12509MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12509MI Supplier MilliporeSigma Supplier No. 12509

Goat Polyclonal Antibody

Goat anti-Mouse IgG (H+L) Antibody, Rhodamine conjugate detects level of Mouse IgG (H+L) and has been published and validated for use in flow cytometry, immunofluoresence, immunocytochemistry, immunohistochemistry.

Specifications

Antigen Goat anti-Mouse IgG (HL)
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Rhodamine
Formulation 0.02M potassium phosphatase, pH 7.2, 150mM NaCl, 0.01% sodium azide and 10mg/mL BSA
Host Species Goat
Quantity 500 μg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Lyophilized: 2 years at 4°C; Rehydrated: 12 months at −20°C.

For Research Use Only