missing translation for 'onlineSavingsMsg'
Learn More

IgG Goat anti-Mouse, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP124PMI
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP124PMI 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP124PMI Supplier MilliporeSigma Supplier No. AP124P

Goat Polyclonal Antibody

Detect Mouse IgG using this Goat anti-Mouse IgG Antibody, Peroxidase Conjugated, H+L validated for use in ELISA, immunohistochemistry and western blotting.

Specifications

Antigen Goat anti-Mouse IgG
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate HRP
Formulation Purified goat IgG conjugated to horseradish peroxidase in buffer containing 0.01M Phosphate Buffered Saline, pH 7.1 with 15mg/mL BSA and 0.01% Thimerosal. Lyophilized.
Host Species Goat
Purification Method Affinity Purified
Quantity 2 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C for up to 12 months
Isotype IgG
Show More Show Less

For Research Use Only