Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 555
Description
To minimize cross-reactivity, the goat anti-mouse IgG whole antibodies have been pre cross-adsorbed against bovine IgG, goat IgG, rabbit IgG, rat IgG, human IgG, and human serum. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in less background staining and cross-reactivity. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. Further passages through additional columns result in highly cross-adsorbed preparations of secondary antibody. The benefits of these extra steps are apparent in multiplexing/multicolor-staining experiments where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically.
Specifications
Specifications
| Antigen | Mouse IgG (H+L) Highly Cross-Adsorbed |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Alexa Fluor Plus 555 |
| Formulation | Proprietary buffer with 0.016% Methylisothiazolone, 0.016% Bromonitrodioxane; pH 6.5 |
| Host Species | Goat |
| Immunogen | Gamma Immunoglobins Heavy and Light chains. |
| Purification Method | Affinity chromatography |
| Quantity | 1 mg |
| Show More |
Frequently Asked Questions (FAQs)
All Alexa Fluor Plus secondary antibodies are compatible with the SlowFade Diamond and ProLong Diamond Antifade Mountants.
Because they are brighter, you would most likely be able to use a lower final concentration of the Alexa Fluor Plus secondary antibodies relative to the original Alexa Fluor secondary antibodies.
Compared to existing Alexa Fluor secondary antibodies, the new Alexa Fluor Plus secondary antibodies use a new proprietary dye chemistry coupled to highly cross-adsorbed antibodies, offering higher sensitivity and signal-to-noise ratios.